Corning™ Microarray Slide Holders Available on GSA/VA Contract for Federal Government customers only.

Plastic containers function as storage boxes for printed arrays

Manufacturer: Corning™ 40081

 View more versions of this product

Catalog No. 07-201-629

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Product Type 25 Slide Holder
    Color Orange
    Material Plastic
    Applications Function as storage boxes for printed arrays
    Quantity 20 Case

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit