Corning™ Microvolume Pipet Tips Available on GSA/VA Contract for Federal Government customers only.

Provide accurate, reliable performance in the stated volume range

Manufacturer: Corning™ 4901

 View more versions of this product

Catalog No. 07-200-526

  Check Availability
Add to Cart

    Specifications

    Product Type Microvolume Tip
    Material Polypropylene
    For Use With (Equipment) Eppendorf and Other Ultramicro Pipetters
    Sterility Nonsterile
    Tip Style Microvolume
    Packaging Bag, 10000
    Quantity 10,000
    Certifications/Compliance DNase, RNase, pyrogen free
    Volume (Metric) 0.5 to 10μL
    Color Natural

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit