Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Centrifuge Tube Top Vacuum Filters Available on GSA/VA Contract for Federal Government customers only.

Filters directly into centrifuge tube minimizing unnecessary transfers. Corning™ Centrifuge Tube Top Filters offer a viewing slot in base that lets you measure volumes as low as 5mL.

Manufacturer:  Corning™ 430314

 View more versions of this product

Catalog No. 09-761-35


Add to cart

Description

Description

Saves time and minimizes costs and error by letting you filter samples directly into a centrifuge tube.

  • Sterile 50mL funnel with filter membrane attached to a 50mL centrifuge tube
  • Eliminates open-air liquid transfer step and extra storage vessels
  • Smaller volumes mean less media waste
  • Funnel is made of clear polystyrene
  • Centrifuge tube is polypropylene, with easy-to-read graduations
  • Filter membranes are low protein binding cellulose acetate
  • Side arm fitting permits use with differential pressure of 5 to 10psi

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Sterile
12 Case
Hose
13.6cm2
Yes
50mL polystyrene filter funnel, 50mL polypropylene receiver centrifuge tube, two centrifuge tube stands, orange HDPE plug seal
50mL
0.45μm
Clear
Tube Top Vacuum Filter System
For the filtration of cell culture media, biological fluids, and other aqueous solutions
4.5cm bottle neck
Cellulose Acetate
Documents
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit