Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ 2.5S Nerve Growth Factor (NGF), Mouse Natural

Designed specifically for use in basic cell biology research

Specifications

Cross Reactivity Mouse
Source Mouse Submaxillary Glands
Type Nerve Growth Factor
For Use With (Application) Useful for the maintenance and differentiation of sympathetic and sensory neurons, as well as neuronal cells in-vitro
Lyophilized Yes
View More Specs

Alert:

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.


Products 2
Catalog Number Mfr. No. Formulation Molecular Weight Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Molecular Weight Quantity Price Quantity & Availability  
CB-40005
View Documents
Corning™
354005
Lyophilized from phosphate buffered saline, carrier free, reconstitute in serum containing medium or aqueous buffer containing BSA 2% 26 10μg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
CB-40005A
View Documents
Corning™
356004
Lyophilized from phosphate buffered saline, carrier free, reconstitute in serum containing medium or aqueous buffer containing BSA 2% 26 100μg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

  • NGFs are isolated from mouse submaxillary glands
  • 2.5S form consists of two β subunits
  • 7S form consists of two α, one β and two Υ subunits
  • Products are tested for biological activity in appropriate assays, such as the ability of growth factors to stimulate proliferation or DNA synthesis
  • Lymphokines and growth factors are also sterile filtered generally using a 0.2µm membrane
  • Tested for the presence of bacteria, fungi and mycoplasma

Specification sheets include detailed information on quality control testing, product source, storage, stability and recommendations on proper experimental concentrations.

Order Info

Both supplied in lyophilized form

Specifications

Specifications

Mouse
Nerve Growth Factor
Yes
2° to 8°C
Mouse Submaxillary Glands
Useful for the maintenance and differentiation of sympathetic and sensory neurons, as well as neuronal cells in-vitro
3 Months
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit