Learn More
Corning™ Osteo Assay Surface Microplates Available on GSA/VA Contract for Federal Government customers only.
Capable of supporting the functional properties of osteogenic cells. Corning™ Osteo Assay Surface delivers lot-to-lot consistency, translating to reliable and reproducible results in bone cell assays. The surface also offers a consistent and defined alternative to preparing dentine or bone slices, reducing the variability in assay systems and resulting in more predictable assay readouts.
Manufacturer:  Corning™ 3987
 View more versions of this product
Catalog No. 09-762-100
Add to cart
Description
Description
 Features:
  • 3 dimensional structure mimics in vivo bone and in vitro bone cell assays
  • Inorganic, non-biological
  • Nonreversible lids with condensation rings to reduce contamination
  • Individual alphanumeric codes for well identification
  • Individually wrapped
  • Sterilized by gamma irradiation
Surface is designed for:
  • Direct assessment of osteoclast and osteoblast functional in vitro activity
  • Osteoclast and osteoblast precursor differentiation
  • Co-culture of osteoclast and/or osteoblasts with other cell lines
  • Solution-based quantitative assays
  • Studies related to bone remodeling and pit formation
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Clear
Osteoblast and osteoclast differentiation and functional studies
Polystyrene
3.4mL
Clear
Round
Sterile
1.9cm2
24 well
Osteo assay surface
380 to 570μL
With Lid
24-Well Osteo Assay Surface Multiwell Microplate