Corning™ 384-Well Clear Bottom Black or White Polystyrene Microplates Available on GSA/VA Contract for Federal Government customers only.

Manufactured from medical grade polystyrene for strength. Corning™ 384-Well Clear Bottom Black or White Polystyrene Microplates permit top or bottom analysis using detection microplate readers.

Manufacturer: Corning™ 3711

 View more versions of this product

Catalog No. 07-200-654

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    For Use With (Application) Hydrophobic in nature and binds bio-molecules through passive interactions. Suitable for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions.
    Type EIA/RIA
    Binding Property Medium bind
    Bottom Flat bottom
    Lid No
    Material Polystyrene
    Sterile No
    Volume (Metric) Well 112μL,112μL
    Volume (Metric) Working 20 to 80μL
    Closure No,No
    Color Black
    Applications For fluorescent and luminescent based assays and SPA

     

    • Molded polystyrene with opaque walls and optically clear bottoms
    • Readable from top or bottom for direct microscope viewing and CCD camera-based detectors
    • Opaque black or white walls prevent well to well crosstalk
    • Recommended working volumes of 20 to 80µL; total well volume of 112µL
    • Sterile microplates are sterilized by gamma radiation and certified nonpyrogenic
    • Suited for flourescent and luminescent assays
    • Plate dimensions meet proposed industry standard

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit