Learn More
Corning™ Polystyrene Poly-D-Lysine Coated Microplates Available on GSA/VA Contract for Federal Government customers only.
Promote cell attachment
Manufacturer:  Corning™ 3846
 View more versions of this product
Catalog No. 08-757-469
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Yes
White, Clear
Easy transition into existing automation and readers, reduced evaporation of well contents, enhanced cell attachment, enhance luminescent signals and have low background luminescence and fluorescence
Polystyrene
Poly-D-Lysine
95μL
Clear
Yes
0.06cm2
384-well
Yes
384-Well Poly-D-Lysine Microplate
20 to 80μL
Square