Learn More
Corning™ PTFE-Coated Stir Bars
Ideal for use with Scholar™ PC-171 stirrers
Manufacturer:  Corning™ 409021
 View more versions of this product
Catalog No. 14-513-106
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Ideal for use with Scholar™ PC-171 stirrers
  • PTFE-coated for increased chemical resistance
Specifications
Specifications
PTFE
All PC-200 series hot plates and stirrers
1
8mm
Magnet
0.31 in.
1 in.
Stir bar
25.4mm