Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Corning™ PureCoat™ Amine Flasks
Positively charged surface is ideal for for preparative cell culture, cell-based, and HTS screening assays
Supplier: Corning™ 356728
Description
Corning™ PureCoat™ amine (positively charged) and carboxyl (negatively charged) surfaces provide improved cell attachment, faster cell proliferation, and enhanced recovery post-thaw over standard TC surfaces. These surfaces function with a broad range of primary, transfected, transformed, and fastidious cell types, and have demonstrated utility in serum-reduced or serum-free conditions.Specifications
25.3 oz. | |
750 mL | |
For preparative cell culture, cell-based and HTS screening assays | |
40/Cs. | |
Rectangular | |
Sterile |
Polystyrene | |
Polystyrene | |
Canted | |
5 Pk. | |
Amine | |
Vented |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.
Spot an opportunity for improvement?
Provide Content Correction