Learn More
Corning™ PureCoat™ Amine Flat Bottom Multiwell Plates
Positively charged surface improves attachment. increases proliferation and improved differentiation for a broad range of cells that exhibit poor attachment properties in serum-free or serum-reduced conditions
Manufacturer:  Corning™ 354717
 View more versions of this product
Catalog No. 08-772-159
Add to cart
Alert:
In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.
Description
Description
  • Chemically defined, animal-free
  • Enhances recovery from freeze-thaw
  • Suitable for primary, transfected and fastidious cell lines
  • Maintains cell monolayers during vigorous liquid handling manipulation
Specifications
Specifications
Clear, Clear
96 well
5/Pk.; 5/Cs.
96-Well Amine Multiwell Microplate
Culture of primary cells, transfected, transformed and fastidious cell lines, culture in standard, serum-free or serum-reduced conditions, freeze-thaw recovery of cryopreserved primary cells and cell lines, including division arrested
Polystyrene
Amine
Sterile
Safety and Handling
Safety and Handling
ShelfLife : 18 Months