Learn More
Corning™ PureCoat™ ECM Mimetic Multi-Flask Available on GSA/VA Contract for Federal Government customers only.
Features multi-layers with vented cap
Manufacturer:  Corning™ 356274
 View more versions of this product
Catalog No. 07-201-379
Add to cart
Description
Description
  • Animal component free, synthetic peptide coating covalently attached to cultureware
  • Compatible with wide range of serum-free, xeno-free, and animal-free media formulations
  • Also compatible with broad range of cell dissociation reagents
  • Streamlines cell culture workflow, reducing handling and contamination risk
  • Predictable scale-up and uniform cell growth
  • Non-cytotoxic
  • Sterile
Specifications
  • Flask shape: Rectangular
  • Neck Style: Straight    
  • Surface treatment: Fibronectin-coated
  • Cap material: Polypropylene
  • Available in 6/Pk. or 8/Pk.
  • Cell growth area: 525cm2 and 875cm2
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Compliance
cGMP manufacturing compliant (ISO 9001:2008, ISO 13485:2012)
Specifications
Specifications
3-layer with blue vented PP cap
2 per pack x 4
Rectangular
525cm2
Straight
Collagen