Learn More
Corning™ PureCoat™ Fibronectin Peptide TC-Treated Microplates
Coated with biologically active synthetic animal-free peptides that are covalently linked to a proprietary surface
$346.50 - $375.00
Specifications
Bottom Shape Flat
Shape Rectangular
View More Specs
Includes: Lid
Products 2
Catalog Number Mfr. No. Well Count Price Quantity & Availability  
Catalog Number Mfr. No. Well Count Price Quantity & Availability  
08-774-350
View Documents
Corning™
356240
6
Case of 10 for $346.50
Add to cart
08-774-351
View Documents
Corning™
356241
24
Case of 10 for $375.00
Add to cart
Description
Description
Corning ECM mimetic surfaces and substrates contain biologically active, synthetic, animal-free peptides that have been rationally designed to mimic the cell attachment motifs of native ECM proteins. The peptides enable optimal cell binding and signaling in a broad range of serum-free, xeno-free, and animal-free media formulations, supporting the propagation and differentiation of a range of stem, progenitor, and primary cell types.
Corning PureCoat cultureware is coated with biologically active, synthetic, animal-free peptides that are covalently linked to a proprietary surface to provide a highly consistent, cost-effective alternative to self-coated extracellular peptides. The proprietary covalent linkage orients the peptides for optimal binding and signaling.
Specifications
Specifications
Flat
Rectangular
Safety and Handling
Safety and Handling
ShelfLife : 18 Months