Learn More
Corning™ PVC-Coated Filtering Flask with Tubulation Products for Science Education
Feature PVC coating for longer life and safety.
$301.00
Specifications
Material Borosilicate Glass
View More Specs
Products 1
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
S339241 Products for Science Education
View Documents
Corning™
65340500/EMD
500 mL
Case of 6 for $301.00
Add to cart
Description
Description
  • Quality filtering flasks with side tubulation
  • Tooled necks accept hose/tubing that has an inner dimension of 8mm (0.31 in.)
Specifications
Specifications
Borosilicate Glass