Learn More
PYREX™ VISTA™ Erlenmeyer Flasks
Uniform wall thickness provides the proper balance between mechanical strength and thermal shock resistance.
Supplier: PYREX™ 70980125/EMD
Description
- Narrow mouth
- Approximate graduations in durable blue enamel
- Extra-large marking space
Order Info
Sold in packs of 12.
Compliance
Designed to ASTM™ E-1404, Type 1.
Specifications
Blue Enamel graduations; Top style: Beaded rim | |
Borosilicate Glass, Pyrex Glass | |
No | |
Non-sterile |
125 mL | |
Extra Large Blue Marking | |
Narrow |