Learn More
PYREX VISTA Flat-Bottom Boiling Flasks
Designed for great shock resistance.
$181.00
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
S76104B
![]() |
Corning
70100250/EMD |
250 mL |
Pack of 6 for $181.00
Case of 2 PK for $361.08 |
|
Description
- Full length standard taper 24/40 outer joints
- Short necks
- Wall thicknesses provide balance between thermal expansion and mechanical strength
Compliance
Designed to ASTM E-1403.
Specifications
Borosilicate Glass | |
24/40 |