Learn More
PYREX VISTA Heavy-Wall Filter Flask
With sidearm tubulations and graduated to show approximate capacity.
$118.00 - $304.00
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
S76108A
![]() |
Corning
70340250/EMD |
250 mL |
Pack of 6 for $118.00
Case of 2 PK for $234.61 |
|
S76108C
![]() |
Corning
703401L/EMD |
1000 mL |
Pack of 6 for $304.00
Case of 2 PK for $607.92 |
|
S76108B
![]() |
Corning
70340500/EMD |
500 mL |
Pack of 6 for $148.00
Case of 2 PK for $295.81 |
|
Description
- Neck finish affords a good fit for rubber stoppers
- Tubulation O.D. on sizes up to 33.81 oz. (1000mL) is 0.39 in. (10mm)
Compliance
Designed to ASTM E-1406, Type 2.
Specifications
Borosilicate Glass |