Learn More
PYREX™ VISTA™ Round-Bottom Boiling Flasks
Designed for great shock resistance.
Supplier: PYREX™ 70320100/EMD
Description
- Full length standard taper outer joints
- Short necks
- Manufactured from No. 7740 borosilicate glass
- Wall thicknesses provide balance between thermal expansion and mechanical strength
Compliance
Designed to ASTM™ E-1403.
Specifications
100 mL | |
Short | |
No |
Borosilicate Glass | |
24/40 | |
Round |