Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ RC Syringe Filters Available on GSA/VA Contract for Federal Government customers only.

High burst strength for assured performance

Manufacturer:  Corning™ 431222

 View more versions of this product

Catalog No. 09-754-23


Add to cart

Description

Description

For tissue culture and HPLC applications. Polypropylene housing.

  • Diameters: 4, 15 or 25mm diameters handle volumes ranging from 0.5 to 100mL
  • Regenerated cellulose (RC): Excellent chemical resistance, low protein binding, best DMSO compatibility
  • Pore Sizes: 0.2µm filter for very fine particulate removal
  • RC membranes are certified not to cause extraneous peaks in the UV region in HPLC applications. RC filters are certified noncytotoxic and nonpyrogenic.

    This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

    Specifications

    Specifications

    Syringe Filter
    Regenerated Cellulose (RC)
    For tissue culture and HPLC
    0.5 to 100mL
    Polypropylene
    25mm
    87psi
    150μL
    0.2μm
    Documents
    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit