Learn More
Corning™ RoboFlask™ Cell Culture Vessels with Caps
Stackable to save valuable incubator space
Manufacturer:  Corning™ 3070
 View more versions of this product
Catalog No. 07200179
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Yes
Orange
92.6cm2
Yes
Rectangular
Tissue culture treated
70mL
2.36 oz.
HDPE
For use in automated cell maintenance and assay systems
Straight
Sterile
70mL
PS (Polystyrene)