Learn More
Corning™ Robolids Available on GSA/VA Contract for Federal Government customers only.
For use with automation equipment
Manufacturer:  Corning™ 3090
 View more versions of this product
Catalog No. 07-200-699
Add to cart
Description
Description
Suitable for wide range of standard microplates and blocks
  • Stackable
  • Lids specifically designed for multiple automated application and removal on well plates or blocks without need of new seal
  • No piercing is necessary
  • Avoid well-to-well cross contamination
  • Durable silicone plugs provide organic solvent resiliency and low extractables
  • Adaptable to extended sample storage conditions—exhibit low evaporation under standard conditions from -20 to +37°C (-4 to +98.6°F) with results comparable to aluminum sealing foil
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Order Info
Available in 96- and 384-well formats.
Specifications
Specifications
Non-sterile
Automation equipment
25.0
96-Well Robolid
50