Learn More
Corning™ Robolids with Corner Notch
GREENER_CHOICE
For use with automation equipment
$934.00
Specifications
Sterility Non-sterile
For Use With Automation equipment
No. per Case 50/Cs.
No. per Pack 25/Pk.
View More Specs
Products 1
Catalog Number Mfr. No. Format Price Quantity & Availability  
Catalog Number Mfr. No. Format Price Quantity & Availability  
07-200-699
GREENER_CHOICE
View Documents Promotion Details
Corning™
3090
96-Well Robolid
Case of 50 for $934.00
Add to cart
Description
Description
  • Combines the sealing ability of a storage mat with the rigidity of a plastic lid
  • Designed for repeated application and removal by automation and for preventing short-term evaporation
  • Silicone sealing plugs for organic solvent resistance and low extractables
  • Can be used manually or with automation
  • Order Info
    Available in 96- and 384-well formats.
    Specifications
    Specifications
    Non-sterile
    Automation equipment
    50/Cs.
    25/Pk.