Learn More
Corning™ SF Medium
Based on a proprietary combination of classical media formulations
$339.00 - $466.00
Specifications
Content And Storage | 2° to 8°C |
---|---|
Sterility | Sterile |
pH | 7.2 to 7.6 |
Quantity | 6 x 500 mL |
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
MT40102CV
|
Corning™
40102CV |
SF Medium, With L-Glutamine; without Phenol Red |
Case of 6 for $339.00
|
|
MT40101CV
|
Corning™
40101CV |
SF Medium, With L-Glutamine |
Case of 6 for $466.00
|
|
Description
SF Medium is based on a proprietary classical media formulation. This medium contains a mixture of select trace elements and high molecular weight carbohydrates, extra vitamins, animal-free protein, and a small amount of high quality bovine serum albumin (1.0 g/L). SF Medium supports superior growth and viability over long-term passages in hybridomas, as well as suspension and adherent cell cultures. This formulation contains L-glutamine.
Specifications
2° to 8°C | |
Sterile | |
7.2 to 7.6 | |
6 x 500 mL |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.