Learn More
Corning™ Single-Metric Scale Reusable Graduated Cylinders
SCI_ED
Translucent polypropylene cylinders with raised graduations at primary scale points.
$150.00 - $217.00
Specifications
Material Polypropylene
Certifications/Compliance ISO 6706
Calibration To Contain
Disposable Non-Disposable
Scale Single
View More Specs
Products 4
Catalog Number Mfr. No. dimCapacityMetric Graduations Price Quantity & Availability  
Catalog Number Mfr. No. dimCapacityMetric Graduations Price Quantity & Availability  
S02290A
SCI_ED
View Documents
Corning™
3022P25/EMD
0.5 mL
Case of 12 for $188.00
Add to cart
S02290D
SCI_ED
View Documents
Corning™
3022P250/EMD
2 mL
Case of 6 for $150.00
Add to cart
S02290E
SCI_ED
View Documents
Corning™
3022P500/EMD
5 mL
Case of 6 for $192.40
Add to cart
S02290G
SCI_ED
View Documents
Corning™
3022P2L/EMD
20 mL
Case of 3 for $217.00
Add to cart
Description
Description
  • Hexagonal base
  • Calibrated “To Contain” (TC)
  • Tolerances according to ISO 6706
Exposure to temperatures above 60°C may affect accuracy.
Specifications
Specifications
Polypropylene
To Contain
Single
Hexagonal
ISO 6706
Non-Disposable
60°C
Graduated Cylinder