Learn More
Corning™ Snap-Seal Disposable Plastic Sample Containers Available on GSA/VA Contract for Federal Government customers only.
Designed for a wide variety of applications and provide a reliable leak-tight seal when closed properly
Manufacturer:  Corning™ 17305X
 View more versions of this product
Catalog No. 02-540-13
Add to cart
Description
Description
  • Convenient one-handed operation
  • Manufactured from recyclable, translucent polypropylene with straight sides and flat bottoms
  • Snap-Seal locking device keeps the cap closed and secure
  • Polypropylene
  • Ideal for liquids or solids
  • Graduated in milliliters and ounces
  • Hinged cap with rough surface for marking with pen or pencil
  • Patented sealing mechanism provides leak-resistant seal
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Snap
Hinged
13mL
16mm
Yes
Disposable Plastic; Translucent Polypropylene
0.45 oz.
Translucent
94mm
0.45 oz., 13mL