Learn More
Corning Surface Modified Stripwell Microplates
Four surface chemistries suit every type of assay
$1157.00
Specifications
For Use With (Application) | In-vitro diagnostic assays |
---|---|
Color | Clear |
Format | Strips |
Material | Polystyrene |
Sterility | Non-sterile |
Catalog Number | Mfr. No. | Surface Treatment | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Surface Treatment | Price | Quantity & Availability | |||||
07-200-585
|
Corning
2506 |
DNA-Bind |
Case of 50 for $1,157.00
|
|
Description
Corning Stripwell microplates may be used in a variety of laboratory assays. The flat bottom strips are designed to easily break apart and are pre-assembled in an “egg-crate” style strip holder that allows each individual well to be positioned back into the microplate once broken.
- Stripwell microplates have 96-well flat bottom polystyrene format.
- Low volume and standard Stripwell microplates have well volumes of 190 µL and 360 µL, respectively.
- 1 x 8 strips are designed to fit only one way into the strip holder, eliminating the chance of misorientation.
Specifications
In-vitro diagnostic assays | |
Strips | |
Non-sterile | |
75 to 200 μL | |
Round |
Clear | |
Polystyrene | |
360 μL | |
96 |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.