Corning™ Synthemax™ Surface Microplates

Unique synthetic surface coated

Manufacturer: Corning™ 3877

 View more versions of this product

Catalog No. 07-201-248

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Lid Yes
    Culture Area 9.5cm2
    For Use With (Application) Surface supports the expansion of stem cells in their undifferentiated state and enables directed differentiation into specialized cell types
    Format 6-well
    Material Polystyrene
    Quantity 12/Cs.
    Sterile Yes
    Storage Requirements Room Temperature
    Surface Coating Synthemax™-T,Synthemax™-T
    Type 6-Well Synthemax™-T Surface Multiwell Microplate
    Volume (Metric) Well 16.8mL,16.8mL
    Volume (Metric) Working 1.9 to 2.9mL
    Well Design Round
    Closure Yes,Yes
    Color Clear
    Applications Surface supports the expansion of stem cells in their undifferentiated state and enables directed differentiation into specialized cell types
    Well Shape Round
    Bottom Plate Material Polystyrene

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit