Learn More
Corning Step-R Repeating Pipettor Syringe Tips, Non-sterile
Corning Step-R Repeating Pipettor Syringe Tips, Non-sterile provide precise, positive-displacement dispensing of viscous or volatile liquids.
Manufacturer: Corning 6625
Description
- Ideal for viscous, dense and volatile liquides
- Minimum increments printed on the syringe itself
- Accurately deliver selected dose
- Made from high quality polypropylene and polyethylene
Specifications
Dispensing of viscous and volatile liquids | |
Non-sterile | |
ISO8655 | |
100/Pk. | |
12.5 mL |
Polypropylene | |
Autoclavable | |
100/Cs. | |
12.5 mL |