Learn More
Corning™ Thermowell™ Gold 96-Well Polypropylene PCR Microplates
Available in full, half, and elevated skirt styles
Manufacturer:  Corning™ 3751
 View more versions of this product
Catalog No. 07-201-214
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
·    With alphanumeric grid for easy sample identification
·    Uniform wall thickness ensures efficient heat transfer
·    Certified DNase- and RNase-free
·    Nonsterile; without lids
·    Automation compatible
Documents
Accessories