Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Ultralow Attachment Flasks

Features a covalently bound hydrogel layer that minimizes cell attachment, protein absorption and cellular activation. Corning™ Ultralow Attachment Flasks have an ergonomic shape that reduces the number of corners, improves cell scraping and allows the use of a larger pipet.

Manufacturer:  Corning™ 3814CONV

 View more versions of this product

Catalog No. 07-200-875

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »

Disclaimers:

Not tested for endotoxin.


Specifications

Description

Vent caps contain a 0.2µm nonwettable membrane sealed to the cap, providing consistent, sterile gas exchange while minimizing the risk of contamination. These caps are highly recommended for use in all CO2 incubators, especially for long-term use.

 

  • Unique covalently bonded hydrogel surface is hydrophilic and neutrally charged
  • Surface is designed to maintain cells in a suspended, unattached state. Greatly reduces binding of attachment and serum proteins, minimizing cell attachment and spreading
  • Prevents stem cells from attachment-mediated differentiation
  • Prevents anchorage-dependent cells from dividing
  • Minimizes protein absorption, enzyme activation, and cellular activation
  • Biologically inert, noncytotoxic and nondegradable
  • Optically clear virgin polystyrene
  • Rectangular flasks have a ramp from the bottom to the canted neck for easier pouring and pipet access and have an anti-tip skirt to enhance stability
  • U-shaped flasks have rounded shoulders for an easier grip and better access when removing or tightening the cap.
  • Lot numbers for ease in traceability
  • 100% integrity tested

 

Description & Specifications

Specifications

0.76 oz.
HDPE
75cm2
Yes
U-shaped
Ultra-low attachment
PS (Polystyrene)
Orange
Vent
For pipetting or scraping
Canted
Sterile
22.5mL
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit