Learn More
Corning™ Universal Fit Hinged Rack Pipet Tips, Nonsterile
GREENER_CHOICE
Hinged closure provides easy stacking and steam penetration during autoclaving
Supplier:  Corning™ 4712
 View more versions of this product
Catalog No. 07-201-762
Add to Cart
Description
Description
  • Corning universal fit tips are designed to provide a reliable fit with most major brand pipettors. (A Pipet Tip Compatibility Guide can be requested or downloaded from the Corning website).
  • Beveled orifice helps ensure accurate fluid delivery.
  • 1-200 μL universal fit tips are graduated at the 250 μL, 500 μL, and 1,000 μL volumes.
  • Select from three packaging options: Stack rack tips feature a stack of five racks, each containing 96 tips, for a total of 480 tips in a space-saving design. Bulk packed tips are nonsterile and economical.
  • RNase-/DNase-free.
  • Nonpyrogenic.
Order Info
Available sterile and nonsterile
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100 to 1000 μL
Yellow
Hinged Rack
Hinged rack
Non-sterile
Hinged closure provides easy stacking and steam penetration during autoclaving
Round
Yes
Product Suggestions
Product Suggestions