Learn More
Corning™ Universal Fit Pipet Tips: Racked, Nonsterile Available on GSA/VA Contract for Federal Government customers only.
Designed to assure a reliable fit with major brand pipetters
Manufacturer:  Corning™ 4867
 View more versions of this product
Catalog No. 07-200-303
Add to cart
Description
Description
Designed to assure a reliable fit with major brand pipetters
  • Beveled orifice helps ensure accurate fluid delivery
  • 1–200µL tips are graduated at 10, 50 and 100µL
  • 100–1000µL tips are graduated at 250, 500, and 1000µL
  • Choice of sterile or nonsterile
  • Racked tips are certified RNase/DNase-free and nonpyrogenic
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Rack
100 to 1000μL
Blue
Round
Universal Fit
Non-sterile
Yes
PP (Polypropylene)