Learn More
Corning™ Universal Fit Pipet Tips: Stack Rack, Nonsterile Available on GSA/VA Contract for Federal Government customers only.
Designed to assure a reliable fit with major brand pipetters
Manufacturer:  Corning™ 4806
 View more versions of this product
Catalog No. 07-200-500
Add to cart
Description
Description
  • Beveled orifice helps ensure accurate fluid delivery
  • 1–200µL tips are graduated at 10, 50 and 100µL
  • Choice of sterile or nonsterile
  • Stack rack tips feature a space-saving design of five racks, with a total of 480 tips
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Rack
1 to 200μL
Natural
10, 50, 100μL
PP (Polypropylene)
Universal Fit
Non-sterile
Yes
Round
Documents
Accessories