Learn More
Corning™ Vascular Endothelial Growth Factor (VEGF), Human Recombinant
Stimulates proliferation of cultured endothelial cells, induces migration of endothelial cells and monocytes
Manufacturer:  Corning™ 354107
Catalog No. CB-40104
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
A broad range of highly purified natural and recombinant lymphokine and growth factor biochemicals designed specifically for use in basic cell biology research.
  • In vivo, VEGF increases permeability of vascular endothelium
  • Supplied lyophilized
  • Products are tested for biological activity in appropriate assays, such as the ability of growth factors to stimulate proliferation or DNA synthesis
  • Lymphokines and growth factors are also sterile filtered generally using a 0.2μm membrane
  • Tested for the presence of bacteria, fungi and mycoplasma
Specification sheets include detailed information on quality control testing, product source, storage, stability and recommendations on proper experimental concentrations.
Specifications
Specifications
Vascular Endothelial Growth Factor
10μg
3 Months
Human
A broad range of highly purified natural and recombinant lymphokine and growth factor biochemicals designed specifically for use in basic cell biology research
Human Recombinant
2° to 8°C