Learn More
Corning™ Vented Screw Caps for Falcon™ Flasks
Designed for use with Falcon 25-, 75-, and 175cm2 flasks
Manufacturer:  Corning™ 354637
 View more versions of this product
Catalog No. 08-774-203
Add to cart
Description
Description
  • Polyethylene
  • Contain a 0.2µm membrane vent that permits consistent gas exchange while preventing passage of bacteria and fungi, reduce the risk of contamination normally associated with standard cell culturing during open incubation
  • Sterile
Specifications
Specifications
Polyethylene
Falcon™ 25cm2 flasks
Cap, Vented
Screw
0.2μm Membrane vent
Sterile