Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Coronavirus Antibody (FIPV3-70), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064754JF646
Description
Coronavirus Monoclonal antibody specifically detects Coronavirus in Virus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)Specifications
Coronavirus | |
Monoclonal | |
Janelia Fluor 646 | |
Coronaviridae | |
Coronavirus cocktail | |
0.125 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG2a |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
FIPV3-70 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction