Learn More
Anti-coronavirus (SARS-CoV-2) spike S2, mouse, mAb, MP Biomedicals™
Monoclonal antibody raised against the coronavirus spike S2 protein, a common target for neutralizing antibodies and vaccines.
Manufacturer:  MP Biomedicals, Inc 08720402
 View more versions of this product
Catalog No. MP08720402
Add to cart
Description
Description
Anti-coronavirus (SARS-CoV-2) spike S2, mouse, mAb. The 2019 Novel Coronavirus (SARS-CoV-2) is a coronavirus identified as the cause of an outbreak of respiratory illness first detected in Wuhan, China. Its genome encodes four structural proteins, i.e. spike (S), envelope (E), membrane (M), and nucleocapsid (N). The spike protein (S-protein) contains two subunits, S1 and S2. S1 defines the range of the hosts and specificity of the virus, thus recognizes and binds with the cell surface receptor. S2 subunit contains basic elements needed for the membrane fusion. The spike protein is the common target for neutralizing antibodies and vaccines.
Specifications
Specifications
Coronavirus Spike S2 Protein
Monoclonal
Mouse
RUO
None
Western Blot
Unconjugated
250 μg
Primary
IgG1
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For research or further manufacturing use only, not for direct human use.