Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coronin-1a Mouse anti-Human, Allophycocyanin/Cy7, Clone: OTI1A5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271680APCCY7

Catalog No. N271680APC7


Add to cart

Description

Description

Coronin-1a Monoclonal antibody specifically detects Coronin-1a in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Coronin-1a
OTI1A5
Flow Cytometry
Affinity Purified, Conjugated
CORO1A
Full length recombinant protein of human Coronin-1a (NP_009005) produced in HEK293T cells.
Immunogen affinity purified
Cell Cycle and Replication, Immunology, Innate Immunity, Neuroscience
Primary
11151.0
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
clipin-A, CLIPINA, CORO1, coronin, actin binding protein, 1A, coronin, actin-binding protein, 1A, coronin-1, coronin-1A, Coronin-like protein A, Coronin-like protein p57, FLJ41407, HCORO1, MGC117380, p57, TACOCLABP, Tryptophan aspartate-containing coat protein
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit