Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coronin 1B Mouse anti-Human, Clone: OTI3D10, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525212

Catalog No. PIMA525212


Add to cart

Description

Description

Members of the coronin family, such as CORO1B, are WD repeat-containing actin-binding proteins that regulate cell motility (Cai et al., 2005 [PubMed 16027158]).
Specifications

Specifications

Coronin 1B
Monoclonal
0.64 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BR76
CORONIN-2, coronin-1B
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
57175
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI3D10
Unconjugated
CORO1B
Liquid
CORO1B
Full length human recombit protein of CORO1B produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit