Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Coronin 3 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518276
Description
This antibody is predicted to react with bovine and porcine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
Coronin 3 is associated with the aggression and metastasis of hepatocellular carcinoma. Coronins are a family of highly evolutionary conserved proteins reportedly involved in the regulation of actin cytoskeletal dynamics, although only coronin 3 has been shown to be related to cancer cell migration.
Specifications
Coronin 3 | |
Polyclonal | |
Unconjugated | |
Coro1c | |
AL022675; AW455561; AW548837; Coro1c; coronin 1c; coronin 3; coronin, actin binding protein 1C; coronin, actin binding protein, 1C; coronin, actin-binding protein, 1C; coronin-1C; Coronin-3; CRN2; CRNN4; hCRNN4; RGD1564490 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
23603 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9ULV4 | |
Coro1c | |
Synthetic peptide sequence (RISKLEQQMAKIAA) corresponding to the C-terminus amino acids of CORO1C (aa 461-474). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction