Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cortactin Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen Cortactin
Applications KnockDown, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB094722
View Documents
Novus Biologicals
NBP303328100UL
100 μg Each for $320.71
Add to cart
 
NB094723
View Documents
Novus Biologicals
NBP30332820UL
20 μg Each for $138.37
Add to cart
 
Description

Description

Cortactin Polyclonal antibody specifically detects Cortactin in Mouse, Rat samples. It is validated for Western Blot, KnockDown
Specifications

Specifications

Cortactin
Unconjugated
Rabbit
Cell Biology, Cellular Markers, Phospho Specific
PBS with 50% glycerol, pH7.3.
2017
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
KnockDown, Western Blot
Purified
RUO
Mouse, Rat
1110020L01Rik, Amplaxin, cortactin, ems1 sequence (mammary tumor and squamous cell carcinoma-associated (p80/85 srcsubstrate), EMS1amplaxin, FLJ34459, Oncogene EMS1, src substrate cortactin
Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human Cortactin (NP_612632.1).
Polyclonal
Affinity purified
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit