Learn More
Drg International IncSupplier Diversity Partner Corticosterone Elisa DFS Item
Manufacturer:  Drg International IncSupplier Diversity Partner EIA4164
An enzyme immunoassay for the quantitative measurement of corticosterone in serum and plasma. Corticosterone is a glucocorticoid secreted by the cortex of the adrenal gland. Corticosterone is produced in response to the stimulation of the adrenal cortex by adrenocorticotropic hormone (ACTH) and is the precursor of aldosterone. Corticosterone is a major indicator of stress since stress increases the production of corticosteroids. enzyme immunoassay for the quantitative measurement of corticosterone in serum and plasma. Corticosterone is a glucocorticoid secreted by the cortex of the adrenal gland. Corticosterone is produced in response to the stimulation of the adrenal cortex by adrenocorticotropic hormone (ACTH) and is the precursor of aldosterone. Corticosterone is a major indicator of stress since stress increases the production of corticosteroids.
Catalog No. 50-194-7605