Learn More
BioVision IncSupplier Diversity Partner Cortisol (human/mouse/rat) ELISA Kit: Cortisol (human/mouse/rat) ELISA Kit: Rapid & convenient colorimetric assay for quantitative measurement of human, mouse, rat cortisol. Sensitivity: 1 ng/ml and detection range: 5 ng/ml to 480 ng/ml. 100 assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K7430100
Cortisol (human/mouse/rat) ELISA Kit:Cortisol is a steroid hormone released from the adrenal cortex in response to the hormone ACTH. It increases blood pressure and blood sugar levels, and suppresses the immune system. Cortisol acts through specific intracellular receptors and has effects on numerous physiologic systems, including immune system, glucose-counter regulation, vascular tone, substrate utilization and bone metabolism. It exists in the blood as either a free form or bound to corticosteroid-binding globulin (CBG). BioVision's Cortisol ELISA kit is a competitive enzyme immunoassay kit for the quantitative measurement of Cortisol in serum and plasma. Cortisol in a sample or a HRP-conjugated Cortisol bound to the wells by immobilized antibody in a competitive manner. The concentration of Cortisol in the samples is then determined by comparing the O.D. of the samples to the standard curve. Sensitivity of the kit is 1 ng/ml and detection range is from 5 ng/ml to 480 ng/ml.
Catalog No. 50-111-9870