Learn More
Cortisone Acetate, Micronized, USP, 97-102%, Spectrum™
Click to view available options
Packaging:
Amber Glass Bottle
C1438, 50-04-4, C23H30O6
Spectrum™ Cortisone Acetate, Micronized, USP is a corticosteroid used to treat many different allergic disorders. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Ignition Residue 0.005
Loss on Drying 0.01
Packaging Amber Glass Bottle
Specific Rotation +208° to +217°