Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cortisone Acetate, Micronized, USP, 97-102%, Spectrum™ Chemical
SDP

Catalog Number 18601862
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
50-04-4
C23H30O6
402.49
MFCD00003609
ITRJWOMZKQRYTA-RFZYENFJSA-N
2-[(1R,3aS,3bS,9aR,9bS,11aS)-1-hydroxy-9a,11a-dimethyl-7,10-dioxo-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]-2-oxoethyl acetate
CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3C(=O)C[C@]12C
50-04-4
C23H30O6
402.49
MFCD00003609
ITRJWOMZKQRYTA-RFZYENFJSA-N
2-[(1R,3aS,3bS,9aR,9bS,11aS)-1-hydroxy-9a,11a-dimethyl-7,10-dioxo-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]-2-oxoethyl acetate
CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3C(=O)C[C@]12C

C1438, 50-04-4, C23H30O6

Spectrum™ Chemical Cortisone Acetate, Micronized, USP is a corticosteroid used to treat many different allergic disorders. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Quantity 100 g
Percent Purity 97 to 102%
Grade USP
Ignition Residue 0.5%
Loss on Drying 1%
Packaging Amber Glass Bottle
Specific Rotation +208° to +217°
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit