Learn More
Cosorbent, Gas Analysis Chemicals, Burrell Scientific
Used for the determination of Carbon Monoxide in gas analysis
Manufacturer:  Burrell Scientific Inc 0397200008
Catalog No. 14-400-152
Add to cart
Description
Description
  • Used for determining Carbon Monoxide in gas analysis
  • Will not absorb Hydrogen, Nitrogen, or Hydrocarbons
  • Must be used in a Bubbler-Pipette
Recommended Storage
Store at room temperature
Warranty and Services
6 Month Limited Warranty
Specifications
Specifications
Cosorbent, Gas Analysis Chemicals
86%, 9%, 2.50%, 2.50%
Liquid
1mm Hg at 145.8°C (295°F)
340°C (644°F)
3°C
7664-93-9, 7732-18-5, 135-19-3, 1317-39-1
290°C (554°F)
1.8
3.4 (Air = 1) at 20°C
Brown
8 oz.