Learn More
Corning™ Costar™ 384-Well Polypropylene Storage Block Available on GSA/VA Contract for Federal Government customers only.
Rigid sidewalls for optimal performance in automated handling
Manufacturer:  Corning™ 3964
 View more versions of this product
Catalog No. 07-200-749
Add to cart
Description
Description
  • Shallow wells for entry by pipet tips
  • Rounded wells for precise liquid handling and reduced wicking
  • Standard ANSI™ footprint
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
ANSI
For high throughput applications requiring added volume
384
Clear
Storage Microplate
Clear
Polypropylene
180μL
Round
Sterile