Learn More
Corning Costar 8-Pette and 12-Pette Multichannel Pipetters
Adjustable volume pipetters can use any number of channels without affecting pipetting accuracy
$1740.00 - $1979.00
Catalog Number | Mfr. No. | Number of Channels | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Number of Channels | Price | Quantity & Availability | |||||
07-200-132
![]() |
Corning
4880 |
12 |
Case for $1,979.00
|
|
07-200-133
![]() |
Corning
4888 |
8 |
Case for $1,740.00
|
|
Description
These20-200 µL multi-channel pipettors feature a unique, ergonomic trigger-style aspiration and dispense control mechanism designed to reduce thumb fatigue during repetitive pipetting.
· Volume adjusts with a vernier-scale spindle
· Pipettors are entirely autoclavable
Order Info
Compatible with all Fisherbrand universal-fit tips.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Autoclavable | |
20 to 200 μL |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.