Learn More
Corning™ Costar™ 8-Pette™ and 12-Pette™ Multichannel Pipetters Available on GSA/VA Contract for Federal Government customers only.
Adjustable volume pipetters can use any number of channels without affecting pipetting accuracy
Manufacturer:  Corning™ 4888
 View more versions of this product
Catalog No. 07-200-133
Add to cart
This item is not returnable. View return policy
Description
Description
  • Ergonomic pistol-grip design eliminates thumb fatigue during repetitive pipetting procedures
  • Micrometer-style volume adjustment spindle allows quick and easy setting in 0.5µL increments from 20 to 200µL
  • Separate tip ejector trigger prevents accidental tip ejection
  • Each unit is inspected and calibrated for accuracy and precision to within ±2%
  • Fully autoclavable
  • Accuracy and precision test results included with every unit
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Order Info
Compatible with all Fisherbrand™ universal-fit tips.
Specifications
Specifications
Yes
8
20 to 200μL