Learn More
Corning™ Clear Polystyrene 96-Well EIA/RIA Microplates Available on GSA/VA Contract for Federal Government customers only.
Manufactured from clear medical grade polystyrene for strength and visibility. Corning™ Clear Polystyrene 96-Well Microplates present an excellent solution for cell culture and colorimetric (absorbance) assays. Use 96-well microplates for laboratory miniaturization and automation.
Manufacturer:  Corning™ 3591
 View more versions of this product
Catalog No. 07-200-36
Add to cart
Description
Description
  • Medical-grade polystyrene
  • Well volume: 320µL to 360µL
  • Working volume: 75µL to 200µL
  • Choice of flat, round, flat Easy Wash™ or V-shaped bottom
  • Treated surface options are not coated, require no special handling and are stable at room temperature
  • Sterile microplates are sterilized by gamma radiation and certified nonpyrogenic
  • Ideal for serial dilutions, hemagglutination or precipitation assays, and general storage applications
  • Plate dimensions meet proposed industry standards
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Medium Binding
Clear
Polystyrene
360μL
Without Lid
EIA/RIA
Flat
Hydrophobic in nature and binds bio-molecules through passive interactions. Suitable for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions.
1 x 50
75 to 200μL
Individually wrapped
Non-sterile