Learn More
Corning™ Microplate Aluminum Sealing Tape Available on GSA/VA Contract for Federal Government customers only.
Provides tight seal to minimize evaporation and condensation. Corning™ 384 Well Microplate Aluminum Sealing Tape enables oil-free operation when used with thermocyclers with heated lids.
Manufacturer:  Corning™ 6569
 View more versions of this product
Catalog No. 07-200-683
Add to cart
Description
Description
  • Easy application and removal for short- and long-term storage
  • Suitable for use between -80°C and 150°C 
  • DMSO-resistant, non-sterile
  • Certified DNase- and RNase-free
  • Prevents sample evaporation
  • Pierceable for access to reaction mix
  • Removes easily without tearing
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Aluminum
Easy application and removal for short and long-term storage, provide tight seal to minimize evaporation and condensation and enables oil-free operation when used with thermo cyclers with heated lids
-80° to 150°C
100
Sealing Tape
Non-sterile
Yes
384-well microplates
100.0